Proteintech
Manufacturer Code:241051AP150UL
Catalog # 50-173-4238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MAGEF1 Fusion Protein Ag21147 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAGE F1 antigen MAGEF1 melanoma antigen family F 1 Melanoma associated antigen F1; MAGE-F1; MAGE-F1 antigen; melanoma antigen family F, 1; melanoma antigen family F1; Melanoma-associated antigen F1
Gene Aliases: MAGEF1
UniProt ID: (Human) Q9HAY2
Entrez Gene ID: (Human) 64110
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.