Invitrogen
Manufacturer Code:PA599775
Catalog # PIPA599775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MAGE2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9630059J11Rik BB131182 HCA3 Hepatocellular carcinoma-associated protein 3 hepatocellular carcinoma-associated protein HCA3 MAGE family member E2 MAGEE2 Mage-e2 MAGE-E2 antigen melanoma antigen family E 2 melanoma antigen family E2 melanoma antigen family E 2 melanoma-associated antigen E2 RGD1564872; Hepatocellular carcinoma-associated protein 3; hepatocellular carcinoma-associated protein HCA3; MAGE-E2 antigen; melanoma antigen family E, 2; melanoma antigen family E2; melanoma antigen, family E, 2; Melanoma-associated antigen E2
Gene Aliases: 9630059J11Rik; BB131182; HCA3; Mage-e2; MAGEE2; RGD1564872
UniProt ID: (Human) Q8TD90
Entrez Gene ID: (Human) 139599, (Rat) 302392, (Mouse) 272790
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.