Novus Biologicals
Manufacturer Code:NBP2831750.1ML
Catalog # NB031104
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human MAGEE2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HCA3hepatocellular carcinoma-associated protein HCA3 Hepatocellular carcinoma-associated protein 3 MAGE-E2 antigen melanoma antigen family E 2 melanoma-associated antigen E2 MGC138154; Hepatocellular carcinoma-associated protein 3; hepatocellular carcinoma-associated protein HCA3; MAGE-E2 antigen; melanoma antigen family E, 2; melanoma antigen family E2; Melanoma-associated antigen E2
Gene Aliases: HCA3; MAGEE2
UniProt ID: (Human) Q8TD90
Entrez Gene ID: (Human) 139599
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.