Abnova
Manufacturer Code:H00057692K
Catalog # 89-204-392
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human MAGEE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human MAGEE1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Alpha-dystrobrevin-associated MAGE Protein; DAMAGE; DAMAGE HCA1 KIAA1587; dystrobrevin-associated MAGE protein; hepatocellular carcinoma-associated HCA1; Hepatocellular carcinoma-associated protein 1; MAGE-E1 antigen; melanoma antigen family E, 1; melanoma antigen family E1; Melanoma-associated antigen E1
Gene Aliases: DAMAGE; HCA1; KIAA1587; MAGEE1
UniProt ID: (Human) Q9HCI5
Entrez Gene ID: (Human) 57692
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.