Novus Biologicals
Manufacturer Code:NBP15683920UL
Catalog # NBP15683920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MAGEB2(melanoma antigen family B 2) The peptide sequence was selected from the N terminal of MAGEB2. Peptide sequence AAAAGVSSTKSKKGAKSHQGEKNASSSQASTSTKSPSEDPLTRKSGSLVQ. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 3.2; Cancer/testis antigen 3.2 cancer/testis antigen family 3 member 2 CT3.2MAGE-B2 antigen DAM6MAGE XP-2 antigen DSS/AHC critical interval MAGE superfamily 6 DSS-AHC critical interval MAGE superfamily 6 MAGE-XP-2 melanoma antigen family B 2 melanoma-associated antigen B2 MGC26438; cancer/testis antigen family 3, member 2; CT3.2; DAM6; DSS-AHC critical interval MAGE superfamily 6; DSS/AHC critical interval MAGE superfamily 6; MAGE XP-2 antigen; MAGE-B2 antigen; melanoma antigen family B, 2; melanoma antigen family B2; Melanoma-associated antigen B2
Gene Aliases: CT3.2; DAM6; MAGE-XP-2; MAGEB2
UniProt ID: (Human) O15479
Entrez Gene ID: (Human) 4113
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.