Novus Biologicals
Manufacturer Code:NBP272575V
Catalog # NBP272575V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F5 |
Immunogen | Full length recombinant protein of human MAGEB18 (NP_775970) produced in HEK293T cells. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: MAGE-B18 antigen; MAGE-B18 antigen melanoma antigen family B 18 melanoma-associated antigen B18 MGC33889; melanoma antigen family B, 18; melanoma antigen family B18; Melanoma-associated antigen B18
Gene Aliases: MAGEB18
UniProt ID: (Human) Q96M61
Entrez Gene ID: (Human) 286514
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.