OriGene
Manufacturer Code:TA800471
Catalog # 50-167-8348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4E12 |
Immunogen | Full length human recombit protein of human MAGEB1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 3.1; cancer/testis antigen family 3, member 1; CT3.1; CT3.1 DAM10 MAGE-Xp MAGEL1; DAM10; DSS-AHC critical interval MAGE superfamily 10; DSS/AHC critical interval MAGE superfamily 10; MAGE-B1 antigen; MAGE-XP antigen; melanoma antigen family B, 1; melanoma antigen family B1; melanoma antigen, family B, 1; Melanoma-associated antigen B1
Gene Aliases: CT3.1; dam1; DAM10; Mage-b1; Mage-rs1; MAGE-Xp; MAGEB1; MAGEL1; MAGEXP; Smage1
UniProt ID: (Human) P43366
Entrez Gene ID: (Dog) 491803, (Human) 4112, (Mouse) 17145
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.