Abnova
Manufacturer Code:H00004099M35J
Catalog # 89-335-083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C7 |
Immunogen | MAG (NP_002352 119 a.a. &sim 208 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody.
Protein Aliases: GMA S-MAG SIGLEC-4A SIGLEC4A; Myelin-associated glycoprotein; sialic acid binding Ig-like lectin 4A; sialic acid-binding immunoglobulin-like lectin 4A; Siglec-4a
Gene Aliases: GMA; MAG; S-MAG; SIGLEC-4A; SIGLEC4A; SPG75
UniProt ID: (Human) P20916
Entrez Gene ID: (Human) 4099
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
membrane-bound signaling molecule
myelin protein
receptor
signaling molecule
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.