Invitrogen
Manufacturer Code:PA530087
Catalog # PIPA530087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 29 and 356 of Human MAG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1B236; Brain neuron cytoplasmic protein 3; GMA S-MAG SIGLEC-4A SIGLEC4A myelin-associated glycoprotein sialic acid binding Ig-like lectin 4A sialic acid-binding immunoglobulin-like lectin 4A; Myelin-associated glycoprotein; S-MAG (C-term.); sialic acid binding Ig-like lectin 4A; Sialic acid-binding Ig-like lectin 4a; sialic acid-binding immunoglobulin-like lectin 4A; Siglec-4a
Gene Aliases: GMA; MAG; S-MAG; SIGLEC-4A; SIGLEC4A; SPG75
UniProt ID: (Human) P20916, (Rat) P07722, (Mouse) P20917
Entrez Gene ID: (Human) 4099, (Rat) 29409, (Mouse) 17136
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
membrane-bound signaling molecule
myelin protein
receptor
signaling molecule
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.