Invitrogen
Manufacturer Code:MA524732
Catalog # PIMA524732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1328CT786.105.125 |
Immunogen | KLH conjugated synthetic peptide of human MAFK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: basic-leucine zipper transcription factor MafK; Erythroid transcription factor NF-E2 p18 subunit; nuclear factor erythroid-2, ubiquitous (p18); Transcription factor MafK; Transcription factor MafK Erythroid transcription factor NF-E2 p18 subunit MAFK; v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog K; v-maf musculoaponeurotic fibrosarcoma oncogene homolog K
Gene Aliases: MAFK; NFE2U; P18
UniProt ID: (Human) O60675
Entrez Gene ID: (Human) 7975
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.