Invitrogen
Manufacturer Code:PA590907
Catalog # PIPA590907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-162 of human MAFG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic leucine zipper transcription factor MafG; hMAF; Transcription factor MafG; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog G; V-maf musculoaponeurotic fibrosarcoma oncogene homolog G
Gene Aliases: hMAF; MAFG
UniProt ID: (Human) O15525
Entrez Gene ID: (Human) 4097
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.