Proteintech
Manufacturer Code:127711AP150UL
Catalog # 50-172-8238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MAFF Fusion Protein Ag3460 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hMafF MAFF Transcription factor MafF U MAF; Transcription factor MafF; U-Maf; v-maf avian musculoaponeurotic fibrosarcoma oncogene family protein F; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog F; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein F; V-maf musculoaponeurotic fibrosarcoma oncogene homolog F
Gene Aliases: hMafF; MAFF; U-MAF
UniProt ID: (Human) Q9ULX9, (Mouse) O54791
Entrez Gene ID: (Human) 23764, (Rat) 366960, (Mouse) 17133
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.