Invitrogen
Manufacturer Code:PA540756
Catalog # PIPA540756
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human MAFB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kreisler; Kreisler (maf-related) leucine zipper protein; Kreisler maf-related leucine zipper homolog; Kreisler maf-related leucine zipper homolog (b-maf); Maf-B; MAFB/Kreisler basic region/leucine zipper transcription factor; Segmentation protein Kr; Transcription factor Maf; Transcription factor Maf-1; Transcription factor MafB; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog B; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein B; V-maf musculoaponeurotic fibrosarcoma oncogene homolog B; V-maf musculoaponeurotic fibrosarcoma oncogene homolog B (avian) KRML MGC43127
Gene Aliases: b-maf; kr; Kreisler; KRML; Krml1; maf; Maf1; MAFB; MCTO; si:ch73-337g9.1
UniProt ID: (Human) B3KNE1, (Rat) P54842, (Mouse) P54841, (Zebrafish) Q98UK4
Entrez Gene ID: (Human) 9935, (Rabbit) 103352307, (Dog) 485873, (Horse) 100055709, (Rat) 54264, (Guinea pig) 100733495, (Mouse) 16658, (Zebrafish) 100537332
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.