Invitrogen
Manufacturer Code:PA579619
Catalog # PIPA579619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human MAF1 recombit protein (Position: R90-E157). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of yeast MAF1; MAF1 negative regulator of RNA polymerase III; Repressor of RNA polymerase III transcription MAF1 homolog; Repressor of RNA polymerase III transcription MAF1 homolog MAF1
Gene Aliases: 1110068E11Rik; AU042856; MAF1
UniProt ID: (Human) Q9H063, (Mouse) Q9D0U6, (Rat) Q5XIH0
Entrez Gene ID: (Human) 84232, (Mouse) 68877, (Rat) 315093
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.