Novus Biologicals
Manufacturer Code:NBP304591100UL
Catalog # NB095041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-256 of human MAF1 (NP_115648.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp586G1123 homolog of yeast MAF1 MAF1 homolog (S. cerevisiae) MGC20332 MGC31779 MGC39758 repressor of RNA polymerase III transcription MAF1 homolog; homolog of yeast MAF1; MAF1 negative regulator of RNA polymerase III; Repressor of RNA polymerase III transcription MAF1 homolog
Gene Aliases: MAF1
UniProt ID: (Human) Q9H063
Entrez Gene ID: (Human) 84232
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.