Abnova
Manufacturer Code:PAB19221
Catalog # 89-120-751
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acids near C-terminus of human MAEL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody rasied against synthetic peptide of MAEL.
For Research Use Only
Protein Aliases: cancer/testis antigen 128; FLJ14904; maelstrom homolog; Protein maelstrom homolog; RP11-102C16.1; spermatogenesis associated 35; testicular tissue protein Li 116
Gene Aliases: CT128; MAEL; SPATA35
UniProt ID: (Human) Q96JY0
Entrez Gene ID: (Human) 84944
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.