Proteintech
Manufacturer Code:266661AP150UL
Catalog # 50-173-5419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MAEL Fusion Protein Ag24524 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer/testis antigen 128; MAEL maelstrom homolog (Drosophila) Protein maelstrom homolog RP11 102C16.1; maelstrom homolog; Protein maelstrom homolog; spermatogenesis associated 35; testicular tissue protein Li 116
Gene Aliases: 4933405K18Rik; AU019877; CT128; MAEL; RGD1309333; SPATA35
UniProt ID: (Human) Q96JY0, (Mouse) Q8BVN9
Entrez Gene ID: (Human) 84944, (Rat) 364039, (Mouse) 98558
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.