Invitrogen
Manufacturer Code:PA519879
Catalog # PIPA519879
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide corresponding to amino acids near the carboxy terminus of human MADD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DENN IG20 RAB3GEP MAP kinase-activating death domain protein Rab3 GDP/GTP exchange factor differentially expressed in normal and neoplastic cells insulinoma glucagonoma clone 20; Differentially expressed in normal and neoplastic cells; Insulinoma glucagonoma clone 20; insulinoma-glucagonoma protein 20; MADD; MAP kinase-activating death domain protein; MAP-kinase activating death domain; Rab3 GDP/GTP exchange factor; Rab3 GDP/GTP exchange protein; Rab3 GEP
Gene Aliases: 9630059K23Rik; DENN; IG20; KIAA0358; MADD; RAB3GEP
UniProt ID: (Human) Q8WXG6, (Mouse) Q80U28, (Rat) O08873
Entrez Gene ID: (Human) 8567, (Mouse) 228355, (Rat) 94193
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.