Invitrogen
Manufacturer Code:PA129177
Catalog # PIPA129177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the second half of human MADD protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Differentially expressed in normal and neoplastic cells; Insulinoma glucagonoma clone 20; MADD; MAP kinase-activating death domain protein; MAP-kinase activating death domain; MAP-Kinase Activating Death Domain DENN Differentially Expressed in Neoplasmic vs. Normal Cells; Rab3 GDP/GTP exchange factor
Gene Aliases: DENN; IG20; KIAA0358; MADD; RAB3GEP
UniProt ID: (Human) Q8WXG6
Entrez Gene ID: (Human) 8567
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.