Novus Biologicals
Manufacturer Code:NBP177033
Catalog # NBP177033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human MADD. The immunogen is located within the last 50 amino acids of MADD. Amino Acid Squence: AAVHSSEEDLRTPPRPVSS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DENNFLJ36300 Differentially expressed in normal and neoplastic cells FLJ35600 IG20 Insulinoma glucagonoma clone 20 insulinoma-glucagonoma protein 20 KIAA0358RAB3GEP MAP kinase-activating death domain protein MAP-kinase activating death domain Rab3 GDP/GTP exchange factor; Differentially expressed in normal and neoplastic cells; Insulinoma glucagonoma clone 20; MADD; MAP kinase-activating death domain protein; MAP-kinase activating death domain; Rab3 GDP/GTP exchange factor
Gene Aliases: DENN; IG20; KIAA0358; MADD; RAB3GEP
UniProt ID: (Human) Q8WXG6
Entrez Gene ID: (Human) 8567
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.