Novus Biologicals
Manufacturer Code:NBP30322420UL
Catalog # NB095397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 110-209 of human MAD4 (NP_006445.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHc12; BHLHC12 bHLHc12Max-interacting transcriptional repressor MAD4 Class C basic helix-loop-helix protein 12 Mad4 homolog MAD4Max-associated protein 4 MAX dimerization protein 4 Max dimerizer 4 MST149 MSTP149 MXD4 MAX dimerization protein 4; Class C basic helix-loop-helix protein 12; Mad4 homolog; Max dimerization protein 4; Max dimerizer 4; Max-associated protein 4; Max-interacting transcriptional repressor MAD4
Gene Aliases: BHLHC12; MAD4; MST149; MSTP149; MXD4
UniProt ID: (Human) Q14582
Entrez Gene ID: (Human) 10608
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.