Novus Biologicals
Manufacturer Code:NBP275562
Catalog # NBP275562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JU99-23 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: hREV7; hREV7 MAD2 mitotic arrest deficient-like 2 (yeast) MAD2BREV7 homolog MAD2-like protein 2 Mitotic arrest deficient 2-like protein 2 mitotic arrest deficient homolog-like 2 mitotic spindle assembly checkpoint protein MAD2B REV7yeast homolog)-like 2; MAD2 (mitotic arrest deficient, yeast, homolog)-like 2; MAD2-like protein 2; Mitotic arrest deficient 2-like protein 2; mitotic arrest deficient homolog-like 2; Mitotic spindle assembly checkpoint protein MAD2B; polymerase (DNA-directed), zeta 2, accessory subunit; REV7 homolog
Gene Aliases: MAD2B; MAD2L2; POLZ2; REV7
UniProt ID: (Human) Q9UI95
Entrez Gene ID: (Human) 10459
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.