Invitrogen
Manufacturer Code:PA575530
Catalog # PIPA575530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 40-85 of Human MAD2L1BP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Caught by MAD2 protein; MAD2L1-binding protein; MAD2L1-binding protein MAD2L1 binding protein Caught by MAD2 protein CMT2 KIAA0110; p31(comet)
Gene Aliases: 0610009D16Rik; 2510031P20Rik; AU018945; AW550977; C76045; CMT2; CMT2A; CMT2B; KIAA0110; MAD2L1BP; Mad2lbp; mKIAA0110
UniProt ID: (Human) Q15013, (Mouse) Q9DCX1
Entrez Gene ID: (Human) 9587, (Mouse) 66591, (Rat) 316237
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.