Invitrogen
Manufacturer Code:PA127838
Catalog # PIPA127838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human MAD2L1-binding protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: Caught by MAD2 protein; CMT2 KIAA0110 MGC11282 RP1-261G23.6; MAD2L1-binding protein; p31(comet)
Gene Aliases: CMT2; KIAA0110; MAD2L1BP
UniProt ID: (Human) Q15013
Entrez Gene ID: (Human) 9587
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.