Proteintech
Manufacturer Code:183221AP150UL
Catalog # 50-173-2113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MAD1 Fusion Protein Ag13171 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hMAD1 HsMAD1 MAD1 MAD1 like protein 1 MAD1L1 PIG9 Tax binding protein 181 TP53I9 TXBP181; HsMAD1; MAD1 mitotic arrest deficient-like 1; MAD1-like protein 1; mitotic arrest deficient 1-like 1; Mitotic arrest deficient 1-like protein 1; Mitotic checkpoint MAD1 protein homolog; Mitotic spindle assembly checkpoint protein MAD1; mitotic-arrest deficient 1, yeast, homolog-like 1; Tax-binding protein 181; tumor protein p53 inducible protein 9
Gene Aliases: AI173502; AW550425; MAD1; MAD1L1; PIG9; TP53I9; TXBP181
UniProt ID: (Human) Q9Y6D9, (Mouse) Q9WTX8
Entrez Gene ID: (Human) 8379, (Rat) 680006, (Mouse) 17120
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.