Invitrogen
Manufacturer Code:PA570915
Catalog # PIPA570915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human MACROD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [Protein ADP-ribosylaspartate] hydrolase MACROD1; [Protein ADP-ribosylglutamate] hydrolase; [Protein ADP-ribosylglutamate] hydrolase MACROD1; ADP-ribose glycohydrolase MACROD1; LRP16; MACRO domain containing 1; MACRO domain-containing protein 1; O-acetyl-ADP-ribose deacetylase MACROD1; Protein LRP16
Gene Aliases: AI604841; AW743046; D930010J01Rik; LRP16; MACROD1
UniProt ID: (Human) Q9BQ69, (Bovine) Q2KHU5, (Rat) Q8K4G6, (Mouse) Q922B1
Entrez Gene ID: (Human) 28992, (Bovine) 613568, (Horse) 102149144, (Dog) 612142, (Rat) 246233, (Guinea pig) 100732197, (Mouse) 107227
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.