Novus Biologicals
Manufacturer Code:NBP287102
Catalog # NB034724
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of HUMAN C2orf65 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome 2 open reading frame 65 D6Mm5e FLJ44410 meiosis 1 associated protein; Meiosis 1 arrest protein; meiosis 1 arresting protein; Meiosis 1-arresting protein; Meiosis 1-associated protein; spermatogenesis associated 37; Spermatogenesis-associated protein 37
Gene Aliases: C2orf65; D6Mm5e; M1AP; SPATA37
UniProt ID: (Human) Q8TC57
Entrez Gene ID: (Human) 130951
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.