Invitrogen
Manufacturer Code:MA530594
Catalog # PIMA530594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E2B1 |
Immunogen | Recombit Human FCN1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Collagen/fibrinogen domain-containing protein 1; FCNM; Ficolin-1; Ficolin-A; Ficolin-alpha; M-ficolin
Gene Aliases: FCN1; FCNM
UniProt ID: (Human) O00602
Entrez Gene ID: (Human) 2219
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.