Invitrogen
Manufacturer Code:PA551552
Catalog # PIPA551552
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human M-Ficolin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Collagen/fibrinogen domain-containing protein 1; Ficolin (collagen/fibrinogen domain containing) 1 FCNM; Ficolin-1; Ficolin-A; Ficolin-alpha; M-ficolin
Gene Aliases: FCN1; FCNM
UniProt ID: (Human) O00602
Entrez Gene ID: (Human) 2219
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.