Invitrogen
Manufacturer Code:PA544192
Catalog # PIPA544192
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human FCN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Collagen/fibrinogen domain-containing protein 1; Collagen/fibrinogen domain-containing protein 2; collagenous lectin; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin (collagen/fibrinogen domain containing) 1; Ficolin (collagen/fibrinogen domain containing) 1 FCNM; ficolin B; Ficolin-1; Ficolin-2; Ficolin-A; Ficolin-alpha; Ficolin-B; Ficolin-beta; L-ficolin; M-ficolin
Gene Aliases: FCN1; FCN2; Fcna; FCNB; FCNM
UniProt ID: (Bovine) Q5I2E5, (Pig) Q29042, (Human) O00602, (Mouse) O70165
Entrez Gene ID: (Bovine) 497016, (Pig) 397316, (Human) 2219, (Guinea pig) 100724936, (Rat) 83517, (Mouse) 14133
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.