R&D Systems
Manufacturer Code:AF3268
Catalog # AF3268
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Phosphopeptide containing human M-CSF R/CD115 Y723 site |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD115; CD115 antigen; CD115 CD115 antigen C-FMS colony stimulating factor 1 receptor CSF1R CSF-1-R CSFR EC 2.7.10.1 FMS proto-oncogene FMSFIM2 macrophage colony stimulating factor I receptor macrophage colony-stimulating factor 1 receptor McDonough feline sarcoma viral (v-fms) oncogene homolog MCSFR M-CSFR Proto-oncogene c-Fms; CSF-1 receptor; CSF-1-R; FMS proto-oncogene; macrophage colony stimulating factor I receptor; Macrophage colony-stimulating factor 1 receptor; McDonough feline sarcoma viral (v-fms) oncogene homolog; Proto-oncogene c-Fms
Gene Aliases: C-FMS; CD115; CSF-1R; CSF1R; CSFR; FIM2; FMS; HDLS; M-CSF-R
UniProt ID: (Human) P07333
Entrez Gene ID: (Human) 1436
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.