R&D Systems
Manufacturer Code:FAB329P025
Catalog # FAB329P025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 61708 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human M-CSF R Ile20-Glu512 (Pro54Ala) Accession # P07333.2 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD115; CD115 antigen; CD115 CD115 antigen C-FMS colony stimulating factor 1 receptor CSF1R CSF-1-R CSFR EC 2.7.10.1 FMS proto-oncogene FMSFIM2 macrophage colony stimulating factor I receptor macrophage colony-stimulating factor 1 receptor McDonough feline sarcoma viral (v-fms) oncogene homolog MCSFR M-CSFR Proto-oncogene c-Fms; CSF-1 receptor; CSF-1-R; FMS proto-oncogene; macrophage colony stimulating factor I receptor; Macrophage colony-stimulating factor 1 receptor; McDonough feline sarcoma viral (v-fms) oncogene homolog; Proto-oncogene c-Fms
Gene Aliases: C-FMS; CD115; CSF-1R; CSF1R; CSFR; FIM2; FMS; HDLS; M-CSF-R
UniProt ID: (Human) P07333
Entrez Gene ID: (Human) 1436
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.