Invitrogen
Manufacturer Code:MA523773
Catalog # PIMA523773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 131621 |
Immunogen | E. coli-derived recombit mouse M-CSF Lys33-Glu262 (predicted) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CSF-1; Macrophage colony-stimulating factor 1; MCSF MGC31930 CSF1; osteopetrosis; Processed macrophage colony-stimulating factor 1
Gene Aliases: C87615; Csf1; Csfm; MCSF; op
UniProt ID: (Mouse) P07141
Entrez Gene ID: (Mouse) 12977
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.