Invitrogen
Manufacturer Code:PA542557
Catalog # PIPA542557
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CSF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: colony stimulating factor 1 (macrophage); Colony stimulating factor 1 (macrophage) MCSF MGC31930 CSF-1; CSF-1; Lanimostim; macrophage colony stimulating factor 1; macrophage colony stimulating factor alpha; macrophage colony stimulating factor beta; Macrophage colony-stimulating factor 1; MCSF; MCSF alpha; osteopetrosis; Processed macrophage colony-stimulating factor 1
Gene Aliases: C87615; CSF-1; CSF1; Csfm; MCSF; MCSFBETA; op
UniProt ID: (Human) P09603, (Mouse) P07141
Entrez Gene ID: (Dog) 611795, (Pig) 100513084, (Bovine) 281094, (Human) 1435, (Guinea pig) 100732898, (Mouse) 12977, (Rat) 78965
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.