Novus Biologicals
Manufacturer Code:NBP159425
Catalog # NBP159425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MCSF The peptide sequence was selected from the middle region of MCSF. Peptide sequence SGSVLPLGELEGRRSTRDRRSPAEPEGGPASEGAARPLPRFNSVPLTDTG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: colony stimulating factor 1 (macrophage); colony stimulating factor 1 (macrophage) CSF-1 Lanimostim macrophage colony stimulating factor macrophage colony-stimulating factor 1 M-CSF MCSFlanimostim MGC31930; CSF-1; Lanimostim; macrophage colony stimulating factor 1; Macrophage colony-stimulating factor 1; Processed macrophage colony-stimulating factor 1
Gene Aliases: CSF-1; CSF1; MCSF
UniProt ID: (Human) P09603
Entrez Gene ID: (Human) 1435
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.