Novus Biologicals
Manufacturer Code:NBP299362AF532
Catalog # NB098870
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Lysyl tRNA synthetase. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMTRIB EC 6.1.1 EC 6.1.1.6 KARS2 KIAA0070 KRS lysine tRNA ligase Lysine--tRNA ligase LysRS lysyl-tRNA synthetase; lysine tRNA ligase; Lysine--tRNA ligase; LysRS; Lysyl-tRNA synthetase
Gene Aliases: CMTRIB; DFNB89; KARS; KARS1; KARS2; KIAA0070; KRS
UniProt ID: (Human) Q15046
Entrez Gene ID: (Human) 3735
Molecular Function: RNA binding protein aminoacyl-tRNA synthetase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.