R&D Systems
Manufacturer Code:AF7164
Catalog # AF7164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Lysosomal Pro-X Carboxypeptidase/PRCP Met1-His496 Accession # P42785 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Angiotensinase C; Angiotensinase C EC 3.4.16.2 HUMPCP Lysosomal carboxypeptidase C lysosomal Pro-X carboxypeptidase PCPMGC2202 Proline carboxypeptidase Prolylcarboxypeptidase prolylcarboxypeptidase (angiotensinase C) prolylcarboxypeptidase isoform 1 preproprotein; Lysosomal carboxypeptidase C; Lysosomal Pro-X carboxypeptidase; PRCP; Proline carboxypeptidase; Prolylcarboxypeptidase; prolylcarboxypeptidase (angiotensinase C)
Gene Aliases: HUMPCP; PCP; PRCP
UniProt ID: (Human) P42785
Entrez Gene ID: (Human) 5547
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.