Novus Biologicals
Manufacturer Code:NBP272553C
Catalog # NBP272553C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5B9 |
Immunogen | Full-length protein expressed in 293T cell transfected with human Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C expression vector. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: [histone H3]-trimethyl-L-lysine(9) demethylase 4C; EC 1.14.11 EC 1.14.11.- GASC-1 protein GASC1JmjC domain-containing histone demethylation protein 3C Gene amplified in squamous cell carcinoma 1 protein JHDM3C JMJD2CbA146B14.1 jumonji domain containing 2C Jumonji domain-containing protein 2C KIAA0780FLJ25949 lysine (K)-specific demethylase 4C lysine-specific demethylase 4C; GASC-1 protein; Gene amplified in squamous cell carcinoma 1 protein; JmjC domain-containing histone demethylation protein 3C; Jumonji domain-containing protein 2C; lysine (K)-specific demethylase 4C; Lysine-specific demethylase 4C; tudor domain containing 14C
Gene Aliases: GASC1; JHDM3C; JMJD2C; KDM4C; KIAA0780; TDRD14C
UniProt ID: (Human) Q9H3R0
Entrez Gene ID: (Human) 23081
Molecular Function: transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.