Novus Biologicals
Manufacturer Code:NB11038884H
Catalog # NB11038884H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 475 and 525 of human Jumonji Domain Containing 2C using the numbering given in entry NP_055876.2 |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-trimethyl-L-lysine(9) demethylase 4C; EC 1.14.11 EC 1.14.11.- GASC-1 protein GASC1JmjC domain-containing histone demethylation protein 3C Gene amplified in squamous cell carcinoma 1 protein JHDM3C JMJD2CbA146B14.1 jumonji domain containing 2C Jumonji domain-containing protein 2C KIAA0780FLJ25949 lysine (K)-specific demethylase 4C lysine-specific demethylase 4C; GASC-1 protein; Gene amplified in squamous cell carcinoma 1 protein; JmjC domain-containing histone demethylation protein 3C; Jumonji domain-containing protein 2C; lysine (K)-specific demethylase 4C; Lysine-specific demethylase 4C; tudor domain containing 14C
Gene Aliases: GASC1; JHDM3C; JMJD2C; KDM4C; KIAA0780; TDRD14C
UniProt ID: (Human) Q9H3R0
Entrez Gene ID: (Human) 23081
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.