Novus Biologicals
Manufacturer Code:NBP151678SS
Catalog # NBP151678SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1000000000000 |
Immunogen | Purified recombinant fragment of human JMJD1A expressed in E. coli. [UniProt# Q9Y4C1] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-dimethyl-L-lysine(9) demethylase 3A; EC 1.14.11 EC 1.14.11.- JHDM2AJMJD1AJHMD2A JmjC domain-containing histone demethylation protein 2A JMJD1 jumonji C domain-containing histone demethylase 2A jumonji domain containing 1 jumonji domain containing 1A Jumonji domain-containing protein 1A KIAA0742DKFZp686A24246 lysine (K)-specific demethylase 3A lysine-specific demethylase 3A testis-specific protein A TSGADKFZp686P07111; JmjC domain-containing histone demethylation protein 2A; jumonji C domain-containing histone demethylase 2A; Jumonji domain-containing protein 1A; lysine (K)-specific demethylase 3A; Lysine-specific demethylase 3A; testis-specific protein A
Gene Aliases: JHDM2A; JHMD2A; JMJD1; JMJD1A; KDM3A; KIAA0742; TSGA
UniProt ID: (Human) Q9Y4C1
Entrez Gene ID: (Human) 55818
Molecular Function: transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.