Invitrogen
Manufacturer Code:MA190371
Catalog # PIMA190371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 31F5 |
Immunogen | This was raised against a yeast nucleolar prep and screened by immunocytochemistry and western blotting. Nucleolar prep contains Lys20p protein (as well as Nop1p/fibrillarin). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HCS; homocitrate synthase LYS20; homocitrate synthase LYS21; Homocitrate synthase, cytosolic isozyme; Homocitrate synthase, mitochondrial
Gene Aliases: D1298; D2195; LYS20; LYS21; YDL131W; YDL182W
UniProt ID: (Baker's yeast) P48570, (Baker's yeast) Q12122
Entrez Gene ID: (Baker's yeast) 851346, (Baker's yeast) 851425
Molecular Function: lyase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.