Novus Biologicals
Manufacturer Code:NBP269989
Catalog # NBP269989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence CPPNKPAESVQSNNS from the internal region of the protein sequence according to NP_057051 NP_036543.4 NP_001180360.1 NP_001295226.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
Protein Aliases: EC 3.1.3.48 Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP Lymphoid phosphatase lymphoid-specific protein tyrosine phosphatase LYP Lyp1 LYP2 PEP PEST-domain phosphatase protein tyrosine phosphatase non-receptor type 22 (lymphoid) protein tyrosine phosphatase non-receptor type 8 PTPN8LYP1 tyrosine-protein phosphatase non-receptor type 22; Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP; Lymphoid phosphatase; lymphoid-specific protein tyrosine phosphatase; LyP; PEP; PEST-domain phosphatase; protein tyrosine phosphatase, non-receptor type 22 (lymphoid); protein tyrosine phosphatase, non-receptor type 8; Tyrosine-protein phosphatase non-receptor type 22
Gene Aliases: LYP; LYP1; LYP2; PEP; PTPN22; PTPN8
UniProt ID: (Human) Q9Y2R2
Entrez Gene ID: (Human) 26191
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.