Novus Biologicals
Manufacturer Code:NBP229697
Catalog # NBP229697
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10A6.2 |
Immunogen | KLH conjugated linear peptide corresponding to the C-terminus of Lyn. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.2 FLJ26625 JTK8 tyrosine-protein kinase Lyn v-yes-1 Yamaguchi sarcoma viral related oncogene homolog Yamaguchi sarcoma viral (v-yes-1) related oncogene homolog; Lck/Yes-related novel protein tyrosine kinase; p53Lyn; p56Lyn; Tyrosine-protein kinase Lyn; V-yes-1 Yamaguchi sarcoma viral related oncogene homolog
Gene Aliases: JTK8; LYN; p53Lyn; p56Lyn
UniProt ID: (Human) P07948
Entrez Gene ID: (Human) 4067
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.