Invitrogen
Manufacturer Code:MA529885
Catalog # PIMA529885
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2 |
Immunogen | Recombit Mouse XCL1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AI661682 ATAC LTN Lptn SCM-1 SCM-1a Scyc1; C motif chemokine 1; Cytokine SCM-1; Lymphotactin; Lymphotaxin; small inducible cytokine subfamily C, member 1 (lymphotactin); Small-inducible cytokine C1
Gene Aliases: AI661682; ATAC; Lptn; LTN; SCM-1; SCM-1a; Scyc1; Xcl1
UniProt ID: (Mouse) P47993
Entrez Gene ID: (Mouse) 16963
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.