Invitrogen
Manufacturer Code:PA575654
Catalog # PIPA575654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human Lymphotactin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATAC; C motif chemokine 1; chemokine (C motif) ligand 1; Cytokine SCM-1; Lymphotactin; Lymphotactin ATAC C motif chemokine 1 Cytokine SCM-1 Lymphotaxin SCM-1-alpha SCM-1alpha SCM1alpha SCM-1-alpha Small-inducible cytokine C1 XC chemokine ligand 1 XCL1 LTN SCYC1 Cytokine SCM-1 beta C motif chemokine 2 XC chemokine ligand 2 XCL2 SCYC2 SCM-1-beta SCM-1beta SCM1beta; Lymphotaxin; SCM-1-alpha; single cysteine motif 1a; small inducible cytokine subfamily C, member 1 (lymphotactin); Small-inducible cytokine C1; XC chemokine ligand 1
Gene Aliases: AI661682; ATAC; LPTN; LTN; SCM-1; SCM-1a; SCM1; SCM1A; SCYC1; XCL1
UniProt ID: (Human) P47992, (Rat) P51672, (Mouse) P47993
Entrez Gene ID: (Human) 6375, (Rat) 171371, (Mouse) 16963
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.