Invitrogen
Manufacturer Code:PA122498
Catalog # PIPA122498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to amino acids 1-92 of Rat Lymphotactin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C motif chemokine 1; Cytokine SCM-1; Ltn Scyc1; Lymphotactin; small inducible cytokine subfamily C, member 1 (lymphotactin); Small-inducible cytokine C1
Gene Aliases: Ltn; Scyc1; Xcl1
UniProt ID: (Rat) P51672
Entrez Gene ID: (Rat) 171371
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.