Novus Biologicals
Manufacturer Code:NBP180637
Catalog # NBP180637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GALDRIVEYLVGDGPQNRYALICQQCFSHNGMALKEEFEYIAFRCAYCFFLNPARKTRPQAPRLPEFSFEKRQVVEGSSSVGPLPSGSVLSSDNQFNEESLEHDVLDDNTEQTDDKIPATEQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310011O18Rik; 2310011O18Rik 4921514L11Rik DKFZp686G1442 KIAA1715 LNP Lunapark MGC102862 protein lunapark Ul ulnaless; 4921514L11Rik; Endoplasmic reticulum junction formation protein lunapark; ER junction formation factor lunapark; limb and neural patterns; Lunapark
Gene Aliases: KIAA1715; LNP; LNP1; LNPK; Ul; ulnaless
UniProt ID: (Human) Q9C0E8
Entrez Gene ID: (Human) 80856
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.