Invitrogen
Manufacturer Code:PA569109
Catalog # PIPA569109
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat Lsm10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: U7 snRNA-associated Sm-like protein LSm10; U7 snRNP-specific Sm-like protein LSM10
Gene Aliases: LSM10; MST074; MSTP074
UniProt ID: (Human) Q969L4, (Mouse) Q8QZX5
Entrez Gene ID: (Rabbit) 100355085, (Horse) 100069271, (Bovine) 618089, (Human) 84967, (Dog) 475335, (Mouse) 116748, (Guinea pig) 100730739, (Rat) 366468, (Zebrafish) 100536270
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.