Abnova
Manufacturer Code:PAB12132
Catalog # 89-115-630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 900-1000 of mouse Lrrk2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of Lrrk2.
For Research Use Only
Protein Aliases: 4921513O20Rik; 9330188B09Rik; AW561911; cI-46; D630001M17Rik; Gm927; Leucine-rich repeat serine/threonine-protein kinase 2
Gene Aliases: 4921513O20Rik; 9330188B09Rik; AW561911; cI-46; D630001M17Rik; Gm927; Lrrk2
UniProt ID: (Mouse) Q5S006
Entrez Gene ID: (Mouse) 66725
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.