Abnova
Manufacturer Code:MAB16105
Catalog # 89-998-880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Lise-1 |
Immunogen | Death domain of mouse Lrdd. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1200011D09Rik; AU042446; leucine-rich and death domain containing; Leucine-rich repeat and death domain-containing protein; p53 protein induced, with death domain; p53-induced death domain-containing protein 1; p53-induced protein with a death domain; Pidd; PIDD-C; PIDD-CC; PIDD-N
Gene Aliases: 1200011D09Rik; AU042446; Lrdd; Pidd; Pidd1
UniProt ID: (Mouse) Q9ERV7
Entrez Gene ID: (Mouse) 57913
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.